4,4'''-Bis(2-butyloctyloxy)-p-quaterphenyl ≥98.0% (by HPLC)

Supplier: TCI America

B5320-1G B5320-200MG
TCB5320-1GEA 195 USD
TCB5320-1G TCB5320-200MG
4,4'''-Bis(2-butyloctyloxy)-p-quaterphenyl ≥98.0% (by HPLC)
4,4'''-Bis(2-butyloctyloxy)-p-quaterphenyl
Formula: C₄₈H₆₆O₂
Melting Pt: 163 °C
MDL Number: MFCD02667784
CAS Number: 18434-08-7
EINECS: 242-310-0

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Molar extinction coefficient min. 51000 (CHCl3, 308 to 312nm)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR