1,3-Difluoro-5-propylbenzene ≥98.0% (by GC)

Supplier: TCI America
Warning

D5134-1G D5134-5G
TCD5134-1GEA 65 USD
TCD5134-1G TCD5134-5G
1,3-Difluoro-5-propylbenzene ≥98.0% (by GC)
1,3-Difluoro-5-propylbenzene
Formula: C₉H₁₀F₂
MW: 156.18 g/mol
Boiling Pt: 170 °C
MDL Number: MFCD12405568
CAS Number: 183245-00-3
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR