N,N'-Di(9-phenanthrenyl)-N,N'-diphenylbenzidine ≥98.0% (by total nitrogen basis)

Supplier: TCI America

Synonyms: N,N'-Bis(9-phenanthrenyl)-N,N'-diphenylbenzidine, N4,N4'-Di(phenanthren-9-yl)-N4,N4'-diphenyl-[1,1'-biphenyl]-4,4'-diamine

D5177-1G D5177-200MG
TCD5177-1GEA 200 USD
TCD5177-1G TCD5177-200MG
N,N'-Di(9-phenanthrenyl)-N,N'-diphenylbenzidine ≥98.0% (by total nitrogen basis)
N,N'-Di(9-phenanthrenyl)-N,N'-diphenylbenzidine
Formula: C₅₂H₃₆N₂
MW: 688.87 g/mol
MDL Number: MFCD03093245
CAS Number: 182507-83-1

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (with Total Nitrogen) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR