Azido-PEG11-amine ≥93.0% (by HPLC)

Supplier: TCI America
Warning

A3007-250MG A3007-50MG
TCA3007-250MGEA 665 USD
TCA3007-250MG TCA3007-50MG
Azido-PEG11-amine ≥93.0% (by HPLC)
Azido-PEG11-amine
Formula: C₂₄H₅₀N₄O₁₁
Storage Temperature: Refrigerator
CAS Number: 1800414-71-4

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (HPLC) min. 93.0 area%
Elemental analysis (Nitrogen) 9.00 to 10.50 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR