5,5''-Bis(trimethylstannyl)-2,2':5',2''-terthiophene ≥98.0% (by HPLC)

Supplier: TCI America
Danger

B5690-1G B5690-200MG
TCB5690-1GEA 450 USD
TCB5690-1G TCB5690-200MG
5,5''-Bis(trimethylstannyl)-2,2':5',2''-terthiophene ≥98.0% (by HPLC)
5,5''-Bis(trimethylstannyl)-2,2':5',2''-terthiophene
Formula: C₁₈H₂₄S₃Sn₂
Storage Temperature: Refrigerator
MDL Number: MFCD30489177
CAS Number: 178931-63-0
UN: 3146
ADR: 6.1,I

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 120.0 to 124.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR