2,4-Bis(phenylsulfonyl)phenol ≥98.0% (by HPLC)

Supplier: TCI America
Warning

B5947-25G B5947-5G
TCB5947-25GEA 135 USD
TCB5947-25G TCB5947-5G
2,4-Bis(phenylsulfonyl)phenol ≥98.0% (by HPLC)
2,4-Bis(phenylsulfonyl)phenol
Formula: C₁₈H₁₄O₅S₂
MDL Number: MFCD29072767
CAS Number: 177325-75-6

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 95.0 %
Melting point 156.0 to 160.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR