Chloro(methyl)(phenyl)(vinyl)silane ≥98.0% (by GC)

Supplier: TCI America
Danger

C3434-25G C3434-5G
TCC3434-25GEA 355 USD
TCC3434-25G TCC3434-5G
Chloro(methyl)(phenyl)(vinyl)silane ≥98.0% (by GC)
Chloro(methyl)(phenyl)(vinyl)silane
Formula: C₉H₁₁ClSi
MW: 182.72 g/mol
Boiling Pt: 80 °C
Flash Pt: 72 °C
MDL Number: MFCD00053208
CAS Number: 17306-05-7
EINECS: 241-333-3
UN: 2987
ADR: 8,II

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR