4,5-Dichloro-2-phenyl-3(2H)-pyridazinone ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 4,5-Dichloro-2-phenylpyridazin-3(2H)-one

D5325-1G D5325-5G
TCD5325-1GEA 23 USD
TCD5325-1G TCD5325-5G
4,5-Dichloro-2-phenyl-3(2H)-pyridazinone ≥98.0% (by GC)
4,5-Dichloro-2-phenyl-3(2H)-pyridazinone
Formula: C₁₀H₆Cl₂N₂O
MW: 241.07 g/mol
MDL Number: MFCD00006470
CAS Number: 1698-53-9
EINECS: 216-917-6

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 163.0 to 167.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR