4-([1,1'-Biphenyl]-4-yl)-6-chloro-2-phenylpyrimidine ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
B5802-1G
B5802-5G
TCB5802-1GEA
100
USD
TCB5802-1G
TCB5802-5G
4-([1,1'-Biphenyl]-4-yl)-6-chloro-2-phenylpyrimidine ≥98.0% (by GC)
4-([1,1'-Biphenyl]-4-yl)-6-chloro-2-phenylpyrimidine
Formula:
C₂₂H₁₅ClN₂ |
MDL Number:
MFCD29477698 CAS Number: 1689538-58-6 |
Specification Test Results
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 144.0 to 148.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...