Clevidipine ≥98.0% (by HPLC)

Supplier: TCI America

C3503-1G C3503-250MG
TCC3503-1GEA 172 USD
TCC3503-1G TCC3503-250MG
Clevidipine ≥98.0% (by HPLC)
Clevidipine
Formula: C₂₁H₂₃Cl₂NO₆
Storage Temperature: Refrigerator
MDL Number: MFCD00940070
CAS Number: 167221-71-8
Merck Index: 14,02352

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 96.0 %
Melting point 141.0 to 145.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR