3-Bromo-4'-chloro-1,1'-biphenyl ≥97.0% (by GC)

Supplier: TCI America

B5764-25G B5764-5G
TCB5764-25GEA 220 USD
TCB5764-25G TCB5764-5G
3-Bromo-4'-chloro-1,1'-biphenyl ≥97.0% (by GC)
3-Bromo-4'-chloro-1,1'-biphenyl
Formula: C₁₂H₈BrCl
MW: 267.55 g/mol
MDL Number: MFCD06201387
CAS Number: 164334-69-4
UN: 3151
ADR: 9,II

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR