2-(tert-Butyldimethylsilyl)-1,3-phenylene bis(trifluoromethanesulfonate) ≥95.0% (by GC)

Supplier: TCI America
Danger

Synonyms: 2-(tert-Butyldimethylsilyl)-1,3-phenylene triflate

B5557-1G B5557-200MG
TCB5557-1GEA 272 USD
TCB5557-1G TCB5557-200MG
2-(tert-Butyldimethylsilyl)-1,3-phenylene bis(trifluoromethanesulfonate) ≥95.0% (by GC)
2-(tert-Butyldimethylsilyl)-1,3-phenylene bis(trifluoromethanesulfonate)
Formula: C₁₄H₁₈F₆O₆S₂Si
Storage Temperature: Refrigerator
CAS Number: 1637638-66-4
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR