Chloro-IB-MECA ≥98.0% (by HPLC)

Supplier: TCI America

C3601-10MG C3601-50MG
TCC3601-10MGEA 132 USD
TCC3601-10MG TCC3601-50MG
Chloro-IB-MECA ≥98.0% (by HPLC)
Chloro-IB-MECA
Formula: C₁₈H₁₈ClIN₆O₄
Storage Temperature: Refrigerator
MDL Number: MFCD01861178
CAS Number: 163042-96-4

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 207.0 to 211.0 deg-C
Elemental analysis (Nitrogen) 14.70 to 16.20 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR