N-(3-Chloro-4-fluorophenyl)-7-fluoro-6-nitro-4-quinazolinamine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 4-(3-Chloro-4-fluoroanilino)-7-fluoro-6-nitroquinazoline

C3276-25G C3276-5G
TCC3276-25GEA 438 USD
TCC3276-25G TCC3276-5G
N-(3-Chloro-4-fluorophenyl)-7-fluoro-6-nitro-4-quinazolinamine ≥98.0% (by HPLC, titration analysis)
N-(3-Chloro-4-fluorophenyl)-7-fluoro-6-nitro-4-quinazolinamine
Formula: C₁₄H₇ClF₂N₄O₂
MW: 336.68 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09998825
CAS Number: 162012-67-1

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR