2-[3'-Bromo[1,1'-biphenyl]-3-yl]-4,6-diphenyl-1,3,5-triazine ≥98.0% (by total nitrogen basis)

Supplier: TCI America
Warning

Synonyms: 2-(3'-Bromo-[1,1'-biphenyl]-3-yl)-4,6-diphenyl-1,3,5-triazine

B5895-1G B5895-5G
TCB5895-1GEA 132 USD
TCB5895-1G TCB5895-5G
2-[3'-Bromo[1,1'-biphenyl]-3-yl]-4,6-diphenyl-1,3,5-triazine ≥98.0% (by total nitrogen basis)
2-[3'-Bromo[1,1'-biphenyl]-3-yl]-4,6-diphenyl-1,3,5-triazine
Formula: C₂₇H₁₈BrN₃
MW: 464.37 g/mol
MDL Number: MFCD30730127
CAS Number: 1606981-69-4

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 96.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 204.0 to 208.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR