3'-Bromo[1,1'-biphenyl]-4-carbonitrile ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
B5965-1G
B5965-5G
TCB5965-1GEA
96
USD
TCB5965-1G
TCB5965-5G
3'-Bromo[1,1'-biphenyl]-4-carbonitrile ≥98.0% (by GC)
3'-Bromo[1,1'-biphenyl]-4-carbonitrile
Formula:
C₁₃H₈BrN |
CAS Number:
160521-46-0 |
Specification Test Results
Appearance | White to Light yellow to Dark green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 62.0 to 66.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...