Butyl 2-Methylbutyrate ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: Butyl 2-Methylbutanoate, 2-Methylbutanoic acid Butyl ester, 2-Methylbutyric acid Butyl ester

B6101-100ML B6101-25ML
TCB6101-100MLEA 98 USD
TCB6101-100ML TCB6101-25ML
Butyl 2-Methylbutyrate ≥98.0% (by GC)
Butyl 2-Methylbutyrate
Formula: C₉H₁₈O₂
Boiling Pt: 176 °C
Flash Pt: 60 °C
MDL Number: MFCD00042902
CAS Number: 15706-73-7
EINECS: 239-798-2
UN: 3272
ADR: 3,III

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR