4-(3-Chlorophenyl)-2,6-diphenylpyrimidine ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America
Warning

C3519-1G C3519-5G
TCC3519-1GEA 87 USD
TCC3519-1G TCC3519-5G
4-(3-Chlorophenyl)-2,6-diphenylpyrimidine ≥98.0% (by HPLC, total nitrogen)
4-(3-Chlorophenyl)-2,6-diphenylpyrimidine
Formula: C₂₂H₁₅ClN₂
MW: 342.83 g/mol
MDL Number: MFCD29477687
CAS Number: 1536209-87-6

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 144.0 to 148.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR