Difluoromethyl phenyl sulfide ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: alpha,alpha-Difluorothioanisole, Difluoromethyl Phenyl sulphide, (Difluoromethylthio)benzene

D4938-1G D4938-5G
TCD4938-1GEA 247 USD
TCD4938-1G TCD4938-5G
Difluoromethyl phenyl sulfide ≥98.0% (by GC)
Difluoromethyl phenyl sulfide
Formula: C₇H₆F₂S
Boiling Pt: 63 °C
MDL Number: MFCD16301013
CAS Number: 1535-67-7
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR