Cesium Lead tribromide, low water content ≥98.0% (by titrimetric analysis)
Supplier: TCI America
|
Danger
|
Synonyms:
CsPbBr3
C3569-1G
C3569-5G
TCC3569-1GEA
68
USD
TCC3569-1G
TCC3569-5G
Cesium Lead tribromide, low water content ≥98.0% (by titrimetric analysis)
Cesium Lead tribromide
Formula:
Br₃CsPb Melting Pt: 555 °C |
CAS Number:
15243-48-8 UN: 2291 ADR: 6.1,III |
Specification Test Results
Appearance | Light yellow to Brown powder to crystal |
Purity (Chelometric Titration) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Water | max. 100 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...