2,6-Bis[(2-hydroxyethyl)amino]toluene ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
2,2'-((2-Methyl-1,3-phenylene)bis(azanediyl))diethanol, 2,2'-[(2-Methyl-1,3-phenylene)bis(azanediyl)]diethanol, 2,2'-[(2-Methyl-1,3-phenylene)diimino]diethanol
B5854-5G
B5854-25G
TCB5854-5GEA
23
USD
TCB5854-5G
TCB5854-25G
2,6-Bis[(2-hydroxyethyl)amino]toluene ≥98.0% (by GC, titration analysis)
2,6-Bis[(2-hydroxyethyl)amino]toluene
Formula:
C₁₁H₁₈N₂O₂ MW: 210.28 g/mol |
MDL Number:
MFCD08703311 CAS Number: 149330-25-6 |
Specification Test Results
Appearance | White to Gray to Red powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 116.0 to 120.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...