2'-Bromospiro[fluorene-9,9'-xanthene] ≥97.0% (by GC)

Supplier: TCI America

B5967-1G B5967-5G
TCB5967-1GEA 148 USD
TCB5967-1G TCB5967-5G
2'-Bromospiro[fluorene-9,9'-xanthene] ≥97.0% (by GC)
2'-Bromospiro[fluorene-9,9'-xanthene]
Formula: C₂₅H₁₅BrO
MW: 411.30 g/mol
MDL Number: MFCD31814409
CAS Number: 1477458-14-2

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Melting point 195.0 to 199.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR