Benzyloxytrimethylsilane ≥95.0% (by GC)

Supplier: TCI America
Warning

B5876-25G B5876-5G
TCB5876-25GEA 131 USD
TCB5876-25G TCB5876-5G
Benzyloxytrimethylsilane ≥95.0% (by GC)
Benzyloxytrimethylsilane
Formula: C₁₀H₁₆OSi
Boiling Pt: 212 °C
MDL Number: MFCD00134529
CAS Number: 14642-79-6

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light red clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR