2-Chloro-4,6-bis(9,9-dimethyl-9H-fluoren-2-yl)-1,3,5-triazine ≥98.0% (by total nitrogen basis)

Supplier: TCI America

C3543-1G C3543-5G
TCC3543-1GEA 153 USD
TCC3543-1G TCC3543-5G
2-Chloro-4,6-bis(9,9-dimethyl-9H-fluoren-2-yl)-1,3,5-triazine ≥98.0% (by total nitrogen basis)
2-Chloro-4,6-bis(9,9-dimethyl-9H-fluoren-2-yl)-1,3,5-triazine
Formula: C₃₃H₂₆ClN₃
MDL Number: MFCD30471507
CAS Number: 1459162-69-6

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (with Total Nitrogen) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR