4,4'-Dibromo-4''-(hexyloxy)triphenylamine ≥97.0% (by total nitrogen basis)
Supplier: TCI America
Synonyms:
4-Bromo-N-(4-bromophenyl)-N-[4-(hexyloxy)phenyl]aniline
D5378-1G
D5378-200MG
TCD5378-1GEA
192
USD
TCD5378-1G
TCD5378-200MG
4,4'-Dibromo-4''-(hexyloxy)triphenylamine ≥97.0% (by total nitrogen basis)
4,4'-Dibromo-4''-(hexyloxy)triphenylamine
Formula:
C₂₄H₂₅Br₂NO |
CAS Number:
1456880-51-5 |
Specification Test Results
Appearance | Colorless to Light orange to Yellow clear liquid to slightly cloudy liquid |
Purity (HPLC) | min. 95.0 area% |
Purity (with Total Nitrogen) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...