1-Bromo-9,9'-spirobi[9H-fluorene] ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 1-Bromo-9,9'-spirobifluorene

B6138-1G B6138-5G
TCB6138-1GEA 115 USD
TCB6138-1G TCB6138-5G
1-Bromo-9,9'-spirobi[9H-fluorene] ≥98.0% (by GC)
1-Bromo-9,9'-spirobi[9H-fluorene]
Formula: C₂₅H₁₅Br
MW: 395.30 g/mol
MDL Number: MFCD31556229
CAS Number: 1450933-18-2

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 212.0 to 216.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR