4,4'-Bis(hexyloxy)-3-methylazobenzene ≥95.0% (by HPLC)

Supplier: TCI America

B4596-500MG B4596-5G
TCB4596-500MGEA 323 USD
TCB4596-500MG TCB4596-5G
4,4'-Bis(hexyloxy)-3-methylazobenzene ≥95.0% (by HPLC)
4,4'-Bis(hexyloxy)-3-methylazobenzene
Formula: C₂₅H₃₆N₂O₂
MW: 396.58 g/mol
CAS Number: 1440509-03-4

Order Now

Specification Test Results

Appearance White to Almost white powder to crystaline
Purity (HPLC) min. 95.0 area%
Elemental analysis (Nitrogen) 6.50 to 7.50 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR