4,4'-Bis(trifluoromethyl)-2,2'-bipyridyl ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 4,4'-Bis(trifluoromethyl)-2,2'-bipyridine

B5463-1G B5463-200MG
TCB5463-1GEA 349 USD
TCB5463-1G TCB5463-200MG
4,4'-Bis(trifluoromethyl)-2,2'-bipyridyl ≥98.0% (by GC, titration analysis)
4,4'-Bis(trifluoromethyl)-2,2'-bipyridyl
Formula: C₁₂H₆F₆N₂
MW: 292.18 g/mol
MDL Number: MFCD00233878
CAS Number: 142946-79-0

Order Now

Specification Test Results

Appearance White to Almost white powder to crystaline
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 78.0 to 82.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR