N,N-Bis(4-bromophenyl)-4-fluoroaniline ≥98.0% (by GC)

Supplier: TCI America
Warning

B5637-5G B5637-1G
TCB5637-5GEA 393 USD
TCB5637-5G TCB5637-1G
N,N-Bis(4-bromophenyl)-4-fluoroaniline ≥98.0% (by GC)
N,N-Bis(4-bromophenyl)-4-fluoroaniline
Formula: C₁₈H₁₂Br₂FN
MDL Number: MFCD31618116
CAS Number: 1429194-04-6

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 103.0 to 107.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR