Bis(4-bromophenyl)iodonium trifluoromethanesulfonate ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

Synonyms: Bis(4-bromophenyl)iodonium triflate

B5259-1G B5259-5G
TCB5259-1GEA 120 USD
TCB5259-1G TCB5259-5G
Bis(4-bromophenyl)iodonium trifluoromethanesulfonate ≥98.0% (by HPLC, titration analysis)
Bis(4-bromophenyl)iodonium trifluoromethanesulfonate
Formula: C₁₃H₈Br₂F₃IO₃S
MW: 587.97 g/mol
MDL Number: MFCD21608478
CAS Number: 139139-81-4

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Ion exchange titration) min. 98.0 %
Melting point 210.0 to 214.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR