Bis(2,4,6-trimethylphenyl)iodonium trifluoromethanesulfonate ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

B5260-1G B5260-5G
TCB5260-1GEA 98 USD
TCB5260-1G TCB5260-5G
Bis(2,4,6-trimethylphenyl)iodonium trifluoromethanesulfonate ≥98.0% (by HPLC, titration analysis)
Bis(2,4,6-trimethylphenyl)iodonium trifluoromethanesulfonate
Formula: C₁₉H₂₂F₃IO₃S
MDL Number: MFCD25541454
CAS Number: 139139-80-3

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Ion exchange titration) min. 98.0 %
Melting point 194.0 to 198.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR