1,4-Bis(dicyclohexylphosphonium)butane bis(tetrafluoroborate) ≥95.0% (by titrimetric analysis)

Supplier: TCI America
Danger

Synonyms: DCyPB-FB

B5991-1G B5991-5G
TCB5991-1GEA 105 USD
TCB5991-1G TCB5991-5G
1,4-Bis(dicyclohexylphosphonium)butane bis(tetrafluoroborate) ≥95.0% (by titrimetric analysis)
1,4-Bis(dicyclohexylphosphonium)butane bis(tetrafluoroborate)
Formula: C₂₈H₅₄B₂F₈P₂
MW: 626.29 g/mol
MDL Number: MFCD22666031
CAS Number: 1389309-54-9
UN: 1759
ADR: 8,III

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Iodometric Titration) min. 95.0 %
Melting point 247.0 to 251.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR