4,4'-Dimethyl-2,2'-bipiperidine (mixture of isomers) ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
D4435-1G
D4435-5G
TCD4435-1GEA
184
USD
TCD4435-1G
TCD4435-5G
4,4'-Dimethyl-2,2'-bipiperidine (mixture of isomers) ≥98.0% (by GC, titration analysis)
4,4'-Dimethyl-2,2'-bipiperidine (mixture of isomers)
Formula:
C₁₂H₂₄N₂ Boiling Pt: 75 °C |
CAS Number:
1378896-71-9 |
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % (total of isomer) |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...