5,5'-Dimethyl-2,2'-bipiperidine (mixture of isomers) ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

D4444-1G D4444-5G
TCD4444-1GEA 145 USD
TCD4444-1G TCD4444-5G
5,5'-Dimethyl-2,2'-bipiperidine (mixture of isomers) ≥98.0% (by titrimetric analysis)
5,5'-Dimethyl-2,2'-bipiperidine (mixture of isomers)
Formula: C₁₂H₂₄N₂
MW: 196.34 g/mol
CAS Number: 1378805-84-5

Order Now

Specification Test Results

Appearance White to Almost white powder to lump
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR