4-Bromo-2,6-diethynylpyridine
Supplier: TCI America
|
Warning
|
B4921-1G
B4921-200MG
TCB4921-1GEA
841
USD
TCB4921-1G
TCB4921-200MG
4-Bromo-2,6-diethynylpyridine
4-Bromo-2,6-diethynylpyridine
Formula:
C₉H₄BrN |
CAS Number:
1374038-64-8 |
Specification Test Results
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 113.0 to 117.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...