2-(3-Biphenylyl)amino-9,9-dimethylfluorene

Supplier: TCI America

Synonyms: N-([1,1'-Biphenyl]-3-yl)-9,9-dimethyl-9H-fluoren-2-amine

B5793-1G B5793-5G
TCB5793-1GEA 76 USD
TCB5793-1G TCB5793-5G
2-(3-Biphenylyl)amino-9,9-dimethylfluorene
2-(3-Biphenylyl)amino-9,9-dimethylfluorene
Formula: C₂₇H₂₃N
MW: 361.49 g/mol
MDL Number: MFCD30471917
CAS Number: 1372778-66-9

Order Now

Specification Test Results

Appearance White to Green to Brown powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 135.0 to 139.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR