2-(4-Aminophenyl)benzoxazol-5-amine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 2-(4-Aminophenyl)benzo[d]oxazol-5-amine, 5-Amino-2-(4-aminophenyl)benzoxazole

A3300-25G A3300-5G
TCA3300-25GEA 262 USD
TCA3300-25G TCA3300-5G
2-(4-Aminophenyl)benzoxazol-5-amine ≥98.0% (by HPLC, titration analysis)
2-(4-Aminophenyl)benzoxazol-5-amine
Formula: C₁₃H₁₁N₃O
MW: 225.25 g/mol
MDL Number: MFCD00445970
CAS Number: 13676-47-6

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 229.0 to 233.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR