1,3:2,4-Bis-O-(3,4-dimethylbenzylidene)-D-sorbitol ≥98.0% (by HPLC)

Supplier: TCI America

B5693-25G B5693-5G
TCB5693-25GEA 60 USD
TCB5693-25G TCB5693-5G
1,3:2,4-Bis-O-(3,4-dimethylbenzylidene)-D-sorbitol ≥98.0% (by HPLC)
1,3:2,4-Bis-O-(3,4-dimethylbenzylidene)-D-sorbitol
Formula: C₂₄H₃₀O₆
Melting Pt: 275 °C
MDL Number: MFCD09038711
CAS Number: 135861-56-2

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Specific rotation [a]20/D +45.0 to +51.0 deg (C=1, pyridine)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR