4-Chloro-6-(4-chlorophenyl)-2-phenylpyrimidine ≥98.0% (by GC)

Supplier: TCI America
Warning

C3518-1G C3518-200MG
TCC3518-1GEA 192 USD
TCC3518-1G TCC3518-200MG
4-Chloro-6-(4-chlorophenyl)-2-phenylpyrimidine ≥98.0% (by GC)
4-Chloro-6-(4-chlorophenyl)-2-phenylpyrimidine
Formula: C₁₆H₁₀Cl₂N₂
MDL Number: MFCD30536778
CAS Number: 1354749-13-5

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 125.0 to 129.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR