10-(4-Bromophenyl)-9,9-dimethyl-9,10-dihydroacridine ≥98.0% (by GC)

Supplier: TCI America

B5535-1G B5535-200MG
TCB5535-1GEA 202 USD
TCB5535-1G TCB5535-200MG
10-(4-Bromophenyl)-9,9-dimethyl-9,10-dihydroacridine ≥98.0% (by GC)
10-(4-Bromophenyl)-9,9-dimethyl-9,10-dihydroacridine
Formula: C₂₁H₁₈BrN
MDL Number: MFCD29919158
CAS Number: 1342892-15-2

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 171.0 to 175.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR