4-(Aminomethyl)cyclohexanamine cis- and trans-mixture ≥96.0% (by GC, titration analysis)

Supplier: TCI America
Danger

Synonyms: 1-Amino-4-(aminomethyl)cyclohexane

A3258-1G A3258-5G
TCA3258-1GEA 138 USD
TCA3258-1G TCA3258-5G
4-(Aminomethyl)cyclohexanamine cis- and trans-mixture ≥96.0% (by GC, titration analysis)
4-(Aminomethyl)cyclohexanamine
Formula: C₇H₁₆N₂
MW: 128.22 g/mol
Boiling Pt: 101 °C
MDL Number: MFCD00049306
CAS Number: 13338-82-4
UN: 2735
ADR: 8,II

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 96.0 % (total of isomer)
Purity (Neutralization titration) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR