1-Bromo-9-phenyl-9H-carbazole ≥97.0% (by GC)

Supplier: TCI America
Warning

B5811-1G B5811-5G
TCB5811-1GEA 254 USD
TCB5811-1G TCB5811-5G
1-Bromo-9-phenyl-9H-carbazole ≥97.0% (by GC)
1-Bromo-9-phenyl-9H-carbazole
Formula: C₁₈H₁₂BrN
MW: 322.21 g/mol
MDL Number: MFCD28975084
CAS Number: 1333002-37-1

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 97.0 %
Melting point 73.0 to 77.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR