3-Bromophenanthrene-9,10-dione ≥95.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 3-Bromo-9,10-phenanthrenedione

B6037-1G B6037-5G
TCB6037-1GEA 55 USD
TCB6037-1G TCB6037-5G
3-Bromophenanthrene-9,10-dione ≥95.0% (by GC)
3-Bromophenanthrene-9,10-dione
Formula: C₁₄H₇BrO₂
MW: 287.11 g/mol
MDL Number: MFCD18072876
CAS Number: 13292-05-2

Order Now

Specification Test Results

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR