2-(2-Bromophenyl)-1H-benzimidazole ≥98.0% (by GC, titration analysis)
Supplier: TCI America
B6092-1G
B6092-5G
TCB6092-1GEA
43
USD
TCB6092-1G
TCB6092-5G
2-(2-Bromophenyl)-1H-benzimidazole ≥98.0% (by GC, titration analysis)
2-(2-Bromophenyl)-1H-benzimidazole
Formula:
C₁₃H₉BrN₂ |
MDL Number:
MFCD00449969 CAS Number: 13275-42-8 |
Specification Test Results
Appearance | White to Yellow to Orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 239.0 to 243.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...