2-Bromo-9,9-dimethyl-10-phenyl-9,10-dihydroacridine ≥96.0% (by GC)

Supplier: TCI America

B5534-1G B5534-200MG
TCB5534-1GEA 997 USD
TCB5534-1G TCB5534-200MG
2-Bromo-9,9-dimethyl-10-phenyl-9,10-dihydroacridine ≥96.0% (by GC)
2-Bromo-9,9-dimethyl-10-phenyl-9,10-dihydroacridine
Formula: C₂₁H₁₈BrN
MDL Number: MFCD27980034
CAS Number: 1319720-64-3

Order Now

Specification Test Results

Appearance White to Green to Brown powder to crystal
Purity (GC) min. 96.0 %
Melting point 122.0 to 126.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR