1,6-Bis(trichlorosilyl)hexane ≥95.0% (by GC)

Supplier: TCI America
Danger

B5923-5G B5923-25G
TCB5923-5GEA 106 USD
TCB5923-5G TCB5923-25G
1,6-Bis(trichlorosilyl)hexane ≥95.0% (by GC)
1,6-Bis(trichlorosilyl)hexane
Formula: C₆H₁₂Cl₆Si₂
Boiling Pt: 281 °C
Flash Pt: 75 °C
MDL Number: MFCD00053189
CAS Number: 13083-94-8
EINECS: 235-994-7
UN: 2987
ADR: 8,II

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR