Abafungin ≥95.0% (by HPLC)

Supplier: TCI America

Synonyms: N-[4-[2-(2,4-Dimethylphenoxy)phenyl]-2-thiazolyl]-1,4,5,6-tetrahydro-2-pyrimidinamine, 4-(2-(2,4-Dimethylphenoxy)phenyl)-N-(1,4,5,6-tetrahydropyrimidin-2-yl)thiazol-2-amine, 4-[2-(2,4-Dimethylphenoxy)phenyl]-N-(1,4,5,6-tetrahydropyrimidin-2-yl)thiazol-2-amine

A3213-250MG A3213-50MG
TCA3213-250MGEA 486 USD
TCA3213-250MG TCA3213-50MG
Abafungin ≥95.0% (by HPLC)
Abafungin
Formula: C₂₁H₂₂N₄OS
MW: 378.49 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00942263
CAS Number: 129639-79-8

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 95.0 area%
Melting point 197.0 to 201.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR