(tert-Butyldimethylsilyloxy)malononitrile ≥93.0% (by GC)

Supplier: TCI America
Danger

Synonyms: (tert-Butyldimethylsilyloxy)propanedinitrile, MAC-TBS Reagent

B5370-1G B5370-200MG
TCB5370-1GEA 481 USD
TCB5370-1G TCB5370-200MG
(tert-Butyldimethylsilyloxy)malononitrile ≥93.0% (by GC)
(tert-Butyldimethylsilyloxy)malononitrile
Formula: C₉H₁₆N₂OSi
MW: 196.33 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD27942977
CAS Number: 128302-78-3
UN: 3276
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to Yellow to Green clear liquid
Purity (GC) min. 93.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR