4-Chloro-3-ethyl-1-methyl-1H-pyrazole-5-carboxylic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

C3644-25G C3644-5G
TCC3644-25GEA 159 USD
TCC3644-25G TCC3644-5G
4-Chloro-3-ethyl-1-methyl-1H-pyrazole-5-carboxylic acid ≥98.0% (by GC, titration analysis)
4-Chloro-3-ethyl-1-methyl-1H-pyrazole-5-carboxylic acid
Formula: C₇H₉ClN₂O₂
MW: 188.61 g/mol
MDL Number: MFCD08459057
CAS Number: 127892-62-0

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 162.0 to 166.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR