Bis(2-chloroethyl)phosphoramidic Dichloride ≥98.0% (by GC)

Supplier: TCI America
Danger

B5908-1G B5908-5G
TCB5908-1GEA 114 USD
TCB5908-1G TCB5908-5G
Bis(2-chloroethyl)phosphoramidic Dichloride ≥98.0% (by GC)
Bis(2-chloroethyl)phosphoramidic Dichloride
Formula: C₄H₈Cl₄NOP
MW: 258.89 g/mol
Boiling Pt: 125 °C
MDL Number: MFCD00191796
CAS Number: 127-88-8
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (Argentmetric Titration) min. 97.0 %
Melting point 52.0 to 56.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR